alpha-Bromo-2'-acetonaphthone, 98%
alpha-Bromo-2'-acetonaphthone, 98%
alpha-Bromo-2'-acetonaphthone, 98%
Thermo Scientific Chemicals

alpha-Bromo-2'-acetonaphthone, 98%

CAS: 613-54-7 | C12H9BrO | 249.107 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 147480100
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS613-54-7
IUPAC Name2-bromo-1-(naphthalen-2-yl)ethan-1-one
Molecular FormulaC12H9BrO
InChI KeyYHXHHGDUANVQHE-UHFFFAOYSA-N
SMILESBrCC(=O)C1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Light yellow to pink to brown to purple
Melting point80°C to 85°C
Appearance (Form)Powder or crystals
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only