N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%
N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%
N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%
Thermo Scientific Chemicals

N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%

CAS: 111-18-2 | C10H24N2 | 172.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 147492500
Price (USD)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS111-18-2
IUPAC Name[6-(dimethylamino)hexyl]dimethylamine
Molecular FormulaC10H24N2
InChI KeyTXXWBTOATXBWDR-UHFFFAOYSA-N
SMILESCN(C)CCCCCCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4350 to 1.4370 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only