alpha,alpha'-dichloro-p-xylène, 98 %, Thermo Scientific Chemicals
alpha,alpha'-dichloro-p-xylène, 98 %, Thermo Scientific Chemicals
alpha,alpha'-dichloro-p-xylène, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

alpha,alpha'-dichloro-p-xylène, 98 %, Thermo Scientific Chemicals

CAS: 623-25-6 | C8H8Cl2 | 175.06 g/mol
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
25 g
100 g
500 g
Conditionnement:
Flacon en verre
Référence 147661000
Prix (EUR)
66,60
Each
Quantité:
100 g
Conditionnement:
Flacon en verre
Grand volume ou format personnalisé
Prix (EUR)
66,60
Each
Identifiants chimiques
CAS623-25-6
IUPAC Name1,4-bis(chloromethyl)benzene
Molecular FormulaC8H8Cl2
InChI KeyZZHIDJWUJRKHGX-UHFFFAOYSA-N
SMILESClCC1=CC=C(CCl)C=C1
Voir plus
Melting point98°C to 102°C
GC>=97.5 %
Appearance (Color)White to light yellow
Appearance (Form)Crystalline solid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only