Salicylic Acid, 99+%
Salicylic Acid, 99+%
Salicylic Acid, 99+%
Thermo Scientific Chemicals

Salicylic Acid, 99+%

CAS: 69-72-7 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 147700025
Price (USD)
127.65
Special offer
Online exclusive
Ends: 28-Dec-2026
150.00
Save 22.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
127.65
Special offer
Online exclusive
Ends: 28-Dec-2026
150.00
Save 22.35
Each
Chemical Identifiers
CAS69-72-7
IUPAC Name2-hydroxybenzoic acid
Molecular FormulaC7H6O3
InChI KeyYGSDEFSMJLZEOE-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point158°C to 161°C
Titration with NaOH99.0 to 101.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only