1-Dodecanesulfonic acid, sodium salt, 99+%
1-Dodecanesulfonic acid, sodium salt, 99+%
1-Dodecanesulfonic acid, sodium salt, 99+%
Thermo Scientific Chemicals

1-Dodecanesulfonic acid, sodium salt, 99+%

CAS: 2386-53-0 | C12H25NaO3S | 272.38 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1479000101 gGlass bottle
Catalog number 147900010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2386-53-0
IUPAC Namesodium dodecane-1-sulfonate
Molecular FormulaC12H25NaO3S
InChI KeyDAJSVUQLFFJUSX-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=99.0 %
Appearance (Color)White to cream
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only