Formic acid, 98+%, pure
Formic acid, 98+%, pure
Formic acid, 98+%, pure
Thermo Scientific Chemicals

Formic acid, 98+%, pure

CAS: 64-18-6 | CH2O2 | 46.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Plastic drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 147930100
Price (EUR)
-
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS64-18-6
IUPAC Nameformic acid
Molecular FormulaCH2O2
InChI KeyBDAGIHXWWSANSR-UHFFFAOYSA-N
SMILESOC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
Titration with NaOH>=98.0 %
Color scale=<20 APHA
Refractive index1.3700 to 1.3720 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only