1,1,2,2-Tetrachloroethane, 98.5%
1,1,2,2-Tetrachloroethane, 98.5%
1,1,2,2-Tetrachloroethane, 98.5%
Thermo Scientific Chemicals

1,1,2,2-Tetrachloroethane, 98.5%

CAS: 79-34-5 | C2H2Cl4 | 167.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 147940250
Price (USD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS79-34-5
IUPAC Name1,1,2,2-tetrachloroethane
Molecular FormulaC2H2Cl4
InChI KeyQPFMBZIOSGYJDE-UHFFFAOYSA-N
SMILESClC(Cl)C(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.0 %
Refractive index1.4930 to 1.4950 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only