Tetramethylammonium fluoride tetrahydrate, 98%
Tetramethylammonium fluoride tetrahydrate, 98%
Tetramethylammonium fluoride tetrahydrate, 98%
Thermo Scientific Chemicals

Tetramethylammonium fluoride tetrahydrate, 98%

CAS: 17787-40-5 | C4H12FN·4H2O | 165.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 148070050
Price (USD)
45.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
45.00
Each
Chemical Identifiers
CAS17787-40-5
IUPAC Nametetramethylazanium tetrahydrate fluoride
Molecular FormulaC4H20FNO4
InChI KeyHQFTZNVQVRRDLN-UHFFFAOYSA-M
SMILESO.O.O.O.[F-].C[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Low melting crystalline mass
Infrared spectrumConforms
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only