3-Chloro-1,2-propanediol, 98%
3-Chloro-1,2-propanediol, 98%
3-Chloro-1,2-propanediol, 98%
Thermo Scientific Chemicals

3-Chloro-1,2-propanediol, 98%

CAS: 96-24-2 | C3H7ClO2 | 110.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 148120250
Price (USD)
26.65
Special offer
Online exclusive
Ends: 28-Dec-2026
31.20
Save 4.55
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
26.65
Special offer
Online exclusive
Ends: 28-Dec-2026
31.20
Save 4.55
Each
Chemical Identifiers
CAS96-24-2
IUPAC Name3-chloropropane-1,2-diol
Molecular FormulaC3H7ClO2
InChI KeySSZWWUDQMAHNAQ-UHFFFAOYNA-N
SMILESOCC(O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Liquid
Refractive index1.4780 to 1.4820 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only