Nitromethane, 99%, pure
Nitromethane, 99%, pure
Nitromethane, 99%, pure
Thermo Scientific Chemicals

Nitromethane, 99%, pure

CAS: 75-52-5 | CH3NO2 | 61.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 148510010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS75-52-5
IUPAC Namenitromethane
Molecular FormulaCH3NO2
InChI KeyLYGJENNIWJXYER-UHFFFAOYSA-N
SMILESC[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
Infrared spectrumConforms
Refractive index1.3810 to 1.3830 (20°C, 589 nm)
Appearance (Form)Clear liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only