Benzanilide, 98%
Benzanilide, 98%
Benzanilide, 98%
Thermo Scientific Chemicals

Benzanilide, 98%

CAS: 93-98-1 | C13H11NO | 197.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
1 kg
Packaging:
View all
Glass bottle
Catalog number 148550050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS93-98-1
IUPAC NameN-phenylbenzamide
Molecular FormulaC13H11NO
InChI KeyZVSKZLHKADLHSD-UHFFFAOYSA-N
SMILESO=C(NC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point161°C to 165°C
Appearance (Color)Yellow-green to grey
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only