Allylthiourea, 98%
Allylthiourea, 98%
Allylthiourea, 98%
Thermo Scientific Chemicals

Allylthiourea, 98%

CAS: 109-57-9 | C4H8N2S | 116.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 148800250
Price (USD)
35.65
Special offer
Online exclusive
Ends: 28-Dec-2026
42.10
Save 6.45
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
35.65
Special offer
Online exclusive
Ends: 28-Dec-2026
42.10
Save 6.45
Each
Chemical Identifiers
CAS109-57-9
IUPAC Name(prop-2-en-1-yl)thiourea
Molecular FormulaC4H8N2S
InChI KeyHTKFORQRBXIQHD-UHFFFAOYSA-N
SMILESNC(=S)NCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Melting point70°C to 78°C
Loss on drying=<1.5 % (50°C, 2 hrs)
Appearance (Form)Powder or crystals or granules and/or chunks
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only