3-Heptanol, 98%
3-Heptanol, 98%
3-Heptanol, 98%
Thermo Scientific Chemicals

3-Heptanol, 98%

CAS: 589-82-2 | C7H16O | 116.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 149091000
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS589-82-2
IUPAC Nameheptan-3-ol
Molecular FormulaC7H16O
InChI KeyRZKSECIXORKHQS-UHFFFAOYNA-N
SMILESCCCCC(O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4190 to 1.4220 (20°C, 589 nm)
GC>=97.5 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only