Benzoic acid, 99%, extra pure
Benzoic acid, 99%, extra pure
Benzoic acid, 99%, extra pure
Thermo Scientific Chemicals

Benzoic acid, 99%, extra pure

Benzoic acid, 99%, C7H6O2, CAS Number-65-85-0, benzenecarboxylic acid, benzeneformic acid, benzenemethanoic acid, benzoesaeure gk, benzoesaeure gv, carboxybenzene, dracylic acid, phenylcarboxylic acid, phenylformic acid, retardex, 2.5kg, (20 wt.% soln. in ethanol) clear, 09,92, 249.0 deg.C, 100.0 | CAS: 65-85-0 | C7H6O2 | 122.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
25 kg
Packaging:
View all
Fibre drum
Catalog number 149130250
Price (USD)
-
Quantity:
25 kg
Packaging:
Fibre drum
Request bulk or custom format
Chemical Identifiers
CAS65-85-0
IUPAC Namebenzoic acid
Molecular FormulaC7H6O2
InChI KeyWPYMKLBDIGXBTP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Melting point121°C to 124°C
Titration with NaOH>=98.5 %
Water=<0.5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only