Xanthine, 98%
Xanthine, 98%
Xanthine, 98%
Thermo Scientific Chemicals

Xanthine, 98%

CAS: 69-89-6 | C5H4N4O2 | 152.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 149170050
Price (USD)
44.10
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
44.10
Each
Chemical Identifiers
CAS69-89-6
IUPAC Name2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular FormulaC5H4N4O2
InChI KeyLRFVTYWOQMYALW-UHFFFAOYSA-N
SMILESO=C1NC2=C(NC=N2)C(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
UV spectrophotometry98 to 102 % (lambda max. 278 nm, pH 10)
Appearance of solution(c=1, 1M NaOH) clear colorless to light yellow soln
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only