Xanthine, 98%
Xanthine, 98%
Xanthine, 98%
Thermo Scientific Chemicals

Xanthine, 98%

CAS: 69-89-6 | C5H4N4O2 | 152.11 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 149171000
価格(JPY)
-
見積もりを依頼する
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS69-89-6
IUPAC Name2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular FormulaC5H4N4O2
InChI KeyLRFVTYWOQMYALW-UHFFFAOYSA-N
SMILESO=C1NC2=C(NC=N2)C(=O)N1
さらに表示
UV spectrophotometry98 to 102 % (lambda max. 278 nm, pH 10)
Appearance of solution(c=1, 1M NaOH) clear colorless to light yellow soln
UV spectrophotometrymolar absorptivity 9300 +/- 2%, on dry basis
HPLC>=97.5 %
Loss on drying=<1 % (105°C, 5 hrs)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only