Sodium formaldehydesulfoxylate dihydrate, 98%
Sodium formaldehydesulfoxylate dihydrate, 98%
Sodium formaldehydesulfoxylate dihydrate, 98%
Thermo Scientific Chemicals

Sodium formaldehydesulfoxylate dihydrate, 98%

CAS: 6035-47-8 | CH3NaO3S·2H2O | 154.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 149190010
Price (USD)
72.90
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
72.90
Each
Chemical Identifiers
CAS6035-47-8
IUPAC Namesodium dihydrate sulfinomethanolate
Molecular FormulaCH7NaO5S
InChI KeyVJWVWUHJLZEHOU-UHFFFAOYNA-N
SMILESO.O.[Na+].OS(=O)C[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Zinc (Zn)=<30 ppm
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration Iodometric>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only