N-Chlorosuccinimide, 97%
N-Chlorosuccinimide, 97%
N-Chlorosuccinimide, 97%
Thermo Scientific Chemicals

N-Chlorosuccinimide, 97%

CAS: 128-09-6 | C4H4ClNO2 | 133.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 149251000
Price (USD)
28.65
Online exclusive
31.50
Save 2.85
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
28.65
Online exclusive
31.50
Save 2.85
Each
Chemical Identifiers
CAS128-09-6
IUPAC Name1-chloropyrrolidine-2,5-dione
Molecular FormulaC4H4ClNO2
InChI KeyJRNVZBWKYDBUCA-UHFFFAOYSA-N
SMILESClN1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(5 % in acetone) Clear to light hazy colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only