4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 95%
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 95%
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 95%
Thermo Scientific Chemicals

4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 95%

CAS: 116-63-2 | C10H9NO4S | 239.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 149270250
Price (USD)
40.65
Special offer
Online exclusive
Ends: 28-Dec-2026
47.60
Save 6.95
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 28-Dec-2026
47.60
Save 6.95
Each
Chemical Identifiers
CAS116-63-2
IUPAC Name4-amino-3-hydroxynaphthalene-1-sulfonic acid
Molecular FormulaC10H9NO4S
InChI KeyRXCMFQDTWCCLBL-UHFFFAOYSA-N
SMILESNC1=C2C=CC=CC2=C(C=C1O)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink to light purple
Appearance (Form)Granular powder
Infrared spectrumConforms
Titration with NaOH>=94 %
Sulfated ash=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only