2-Picoline, 98%
2-Picoline, 98%
2-Picoline, 98%
Thermo Scientific Chemicals

2-Picoline, 98%

CAS: 109-06-8 | C6H7N | 93.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 149330010
Price (USD)
74.10
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
74.10
Each
Chemical Identifiers
CAS109-06-8
IUPAC Name2-methylpyridine
Molecular FormulaC6H7N
InChI KeyBSKHPKMHTQYZBB-UHFFFAOYSA-N
SMILESCC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Water=<0.2 %
Refractive index1.4990 to 1.5010 (20°C, 589 nm)
Appearance (Color)Clear light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only