N-Ethylmorpholine, 99%
N-Ethylmorpholine, 99%
N-Ethylmorpholine, 99%
Thermo Scientific Chemicals

N-Ethylmorpholine, 99%

CAS: 100-74-3 | C6H13NO | 115.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
2.5 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 149400025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS100-74-3
IUPAC Name4-ethylmorpholine
Molecular FormulaC6H13NO
InChI KeyHVCNXQOWACZAFN-UHFFFAOYSA-N
SMILESCCN1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4400 to 1.4420 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only