2-Chloroethyl vinyl ether, ca. 95%
2-Chloroethyl vinyl ether, ca. 95%
2-Chloroethyl vinyl ether, ca. 95%
Thermo Scientific Chemicals

2-Chloroethyl vinyl ether, ca. 95%

CAS: 110-75-8 | C4H7ClO | 106.55 g/mol
Have Questions?
Catalog NumberQuantity
149431000100 g
Catalog number 149431000
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS110-75-8
IUPAC Name(2-chloroethoxy)ethene
Molecular FormulaC4H7ClO
InChI KeyDNJRKFKAFWSXSE-UHFFFAOYSA-N
SMILESClCCOC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=94 %
Refractive index1.4370 to 1.4390 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only