Tetraethylene glycol, 99.5%
Tetraethylene glycol, 99.5%
Tetraethylene glycol, 99.5%
Thermo Scientific Chemicals

Tetraethylene glycol, 99.5%

CAS: 112-60-7 | C8H18O5 | 194.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Plastic drum
Catalog number 149590100
Price (USD)
319.65
Special offer
Online exclusive
Ends: 28-Dec-2026
376.00
Save 56.35
Each
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
319.65
Special offer
Online exclusive
Ends: 28-Dec-2026
376.00
Save 56.35
Each
Chemical Identifiers
CAS112-60-7
IUPAC Name2-{2-[2-(2-hydroxyethoxy)ethoxy]ethoxy}ethan-1-ol
Molecular FormulaC8H18O5
InChI KeyUWHCKJMYHZGTIT-UHFFFAOYSA-N
SMILESOCCOCCOCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4581 to 1.4600 (20°C, 589 nm)
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only