DL-1-Amino-2-propanol, 94%, contains approx. 5% 2-Amino-1-propanol
DL-1-Amino-2-propanol, 94%, contains approx. 5% 2-Amino-1-propanol
DL-1-Amino-2-propanol, 94%, contains approx. 5% 2-Amino-1-propanol
Thermo Scientific Chemicals

DL-1-Amino-2-propanol, 94%, contains approx. 5% 2-Amino-1-propanol

CAS: 78-96-6 | C3H9NO | 75.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 149650010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS78-96-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<50 APHA
Infrared spectrumConforms
GC>=93 %
Water=<0.5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only