5-Methyl-2-hexanone, 99%
5-Methyl-2-hexanone, 99%
5-Methyl-2-hexanone, 99%
Thermo Scientific Chemicals

5-Methyl-2-hexanone, 99%

CAS: 110-12-3 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 149660010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS110-12-3
IUPAC Name5-methylhexan-2-one
Molecular FormulaC7H14O
InChI KeyFFWSICBKRCICMR-UHFFFAOYSA-N
SMILESCC(C)CCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Refractive index1.4060 to 1.4090 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only