2-(2-Butoxyethoxy)ethanol, 99+%
2-(2-Butoxyethoxy)ethanol, 99+%
2-(2-Butoxyethoxy)ethanol, 99+%
Thermo Scientific Chemicals

2-(2-Butoxyethoxy)ethanol, 99+%

CAS: 112-34-5 | C8H18O3 | 162.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 149710025
Price (USD)
81.65
Online exclusive
90.60
Save 8.95
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
81.65
Online exclusive
90.60
Save 8.95
Each
Chemical Identifiers
CAS112-34-5
IUPAC Name2-(2-butoxyethoxy)ethan-1-ol
Molecular FormulaC8H18O3
InChI KeyOAYXUHPQHDHDDZ-UHFFFAOYSA-N
SMILESCCCCOCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Specific gravity(20°C) 0.952 to 0.958
Acidity (CH3COOH)=<0.05 %
Infrared spectrumConforms
Water=<0.2 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only