4-Heptanol, 93%
4-Heptanol, 93%
4-Heptanol, 93%
Thermo Scientific Chemicals

4-Heptanol, 93%

CAS: 589-55-9 | C7H16O | 116.2 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1498500505 mLGlass bottle
Catalog number 149850050
Price (USD)
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS589-55-9
IUPAC Nameheptan-4-ol
Molecular FormulaC7H16O
InChI KeyYVBCULSIZWMTFY-UHFFFAOYSA-N
SMILESCCCC(O)CCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=91 %
Appearance (Color)Clear colorless to light yellow
Refractive index1.4175 to 1.4215 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only