DL-3-Methyl-2-butanol, 98%
DL-3-Methyl-2-butanol, 98%
DL-3-Methyl-2-butanol, 98%
Thermo Scientific Chemicals

DL-3-Methyl-2-butanol, 98%

CAS: 598-75-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 149935000
Price (USD)
304.65
Special offer
Online exclusive
Ends: 28-Dec-2026
358.00
Save 53.35
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
304.65
Special offer
Online exclusive
Ends: 28-Dec-2026
358.00
Save 53.35
Each
Chemical Identifiers
CAS598-75-4
IUPAC Name3-methylbutan-2-ol
Molecular FormulaC5H12O
InChI KeyMXLMTQWGSQIYOW-UHFFFAOYNA-N
SMILESCC(C)C(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4080 to 1.4100 (20°C, 589 nm)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only