n-Propyl Ether, 99+%
n-Propyl Ether, 99+%
n-Propyl Ether, 99+%
Thermo Scientific Chemicals

n-Propyl Ether, 99+%

CAS: 111-43-3 | C6H14O | 102.18 g/mol
Have Questions?
Catalog NumberQuantity
15004050050 mL
Catalog number 150040500
Price (USD)
-
Quantity:
50 mL
Request bulk or custom format
Chemical Identifiers
CAS111-43-3
IUPAC Name1-propoxypropane
Molecular FormulaC6H14O
InChI KeyPOLCUAVZOMRGSN-UHFFFAOYSA-N
SMILESCCCOCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=99.0 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Refractive index1.3790 to 1.3810 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only