Orion™ 2100 Series and 1800 Series Analyzer Accessories
Actual product may vary
Thermo Scientific™
Orion™ 2100 Series and 1800 Series Analyzer Accessories
Choose from this assortment of analyzer replacement accessories, such as air pumps, diffusion tubing and flow cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
150060Diffusion Tubing Kit
181170Inlet Sample Filter
Catalog number 150060
Price (USD)
165.60
Online Exclusive
184.00
Save 18.40 (10%)
Each
Type:
Diffusion Tubing Kit
Price (USD)
165.60
Online Exclusive
184.00
Save 18.40 (10%)
Each
Choose from this assortment of Thermo Scientific™ Orion™ 2100 Series and 1800 Series Analyzer Accessories, including various replacement parts such as air pumps, diffusion tubing and flow cells.
Replacement accessories are for use with Orion 2100 series and 1800 series analyzers.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionFor use with acetic acid and ammonia reagents used on 2110XP, 2111XP, 1810AO, 1811EL analyzers
For Use With (Equipment)Thermo Scientific 1800 and 2100 Series Analyzer
Includes4 in. diffusion tubing assembly with O-rings
TypeDiffusion Tubing Kit
Unit SizeEach