Glucose, powder - FAQs
Glucose, powder - FAQs
View additional product information for Glucose, powder - FAQs (15023021)
1 product FAQs found
Is Glucose, powder (Cat. No. 15023021) sterile?
Yes, Glucose, powder (Cat. No. 15023021) is sterile.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.