4-Chlorobenzyl chloride, 99+%
4-Chlorobenzyl chloride, 99+%
4-Chlorobenzyl chloride, 99+%
Thermo Scientific Chemicals

4-Chlorobenzyl chloride, 99+%

CAS: 104-83-6 | C7H6Cl2 | 161.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Catalog number 150240010
Price (USD)
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS104-83-6
IUPAC Name1-chloro-4-(chloromethyl)benzene
Molecular FormulaC7H6Cl2
InChI KeyJQZAEUFPPSRDOP-UHFFFAOYSA-N
SMILESClCC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
GCImpurities profile
GC=<0.5 % p-Chlorotoluene
GC=<0.5 % p-Chlorobenzal chloride
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only