Nunc™ IVF ICSI Dish
Nunc™ IVF ICSI Dish
Nunc™ IVF ICSI Dish
Thermo Scientific™

Nunc™ IVF ICSI Dish

Experience consistently reliable, high-quality results during fertilization. Thermo Scientific™ Nunc™ IVF ICSI Dishes enable clear viewing of samples with a super-polished optical center and meet the exacting requirements of IVF testing, certification, and validation.
Have Questions?
Catalog NumberDiameter (Metric)
15026551 mm
Catalog number 150265
Price (USD)
228.00
Case of 120
Diameter (Metric):
51 mm
Price (USD)
228.00
Case of 120

Experience consistently reliable, high-quality results during fertilization. Thermo Scientific™ Nunc IVF ICSI Dishes meet the exacting requirements of IVF testing, certification, and validation.

Innovative features for quality and ease-of-use

  • Maintain optimal temperature with true flat bottom
  • Low profile provides optimal angles during ICSI process
  • Clearly see sample with super polished optical center
  • Increased safety with easy grip design
  • Certificates of conformity available for every lot

Rigorously tested, certified and validated for IVF

  • CE-marked (Notified Body 2797), FDA-listed
  • Produced under ISO 13485 with full lot traceability
  • Certificates of conformity for your assurance on every lot

Sterile (SAL 10-6)

  • Non-pyrogenic (LAL test)
  • USP Class VI test
  • Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceCE-marked, FDA-cleared, MEA-tested
DescriptionIVF ICSI Dish, Lid w/ Airvent
No. per Case120
No. per Pack1
TypeDish
Bottom ShapeFlat
ColorClear
Diameter (Metric)51 mm
Height (Metric)9 mm
MaterialPolystyrene (USP Class VI)
SterilitySterile
Surface TreatmentUntreated
Well Count1
Unit SizeCase of 120