2,3-Butanedione monoxime, 98%
2,3-Butanedione monoxime, 98%
2,3-Butanedione monoxime, 98%
Thermo Scientific Chemicals

2,3-Butanedione monoxime, 98%

CAS: 57-71-6 | C4H7NO2 | 101.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 150371000
Price (USD)
78.65
Special offer
Online exclusive
Ends: 28-Dec-2026
92.10
Save 13.45
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
78.65
Special offer
Online exclusive
Ends: 28-Dec-2026
92.10
Save 13.45
Each
Chemical Identifiers
CAS57-71-6
IUPAC Name(3E)-3-(hydroxyimino)butan-2-one
Molecular FormulaC4H7NO2
InChI KeyFSEUPUDHEBLWJY-HWKANZROSA-N
SMILESCC(=O)C(\C)=N\O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Sulfated ash=<0.1 %
Solubility(5 % in water) Clear to slightly hazy
Appearance (Color)White to off-white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only