Nunc™ EasYDish™ Cell Culture Dishes, TC Surface, 60 x 16 mm, 21.5 cm2 - FAQs
Nunc™ EasYDish™ Cell Culture Dishes, TC Surface, 60 x 16 mm, 21.5 cm2 - FAQs
View additional product information for Nunc™ EasYDish™ Cell Culture Dishes, TC Surface - FAQs (150468, 150466, 150464, 150462, 150460)
2 product FAQs found
What is a good alternative for Nunc EasYDish Dish, Cat. No. 150466?
Nunc EasYDish Dish, Cat. No. 150464 is the best fit alternative. The only difference is that the height is 17mm, whereas the height of Cat. No. 150466 is 21 mm.
What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?
Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.
The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells
More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .