Ethyl isovalerate, 99%
Ethyl isovalerate, 99%
Ethyl isovalerate, 99%
Thermo Scientific Chemicals

Ethyl isovalerate, 99%

CAS: 108-64-5 | C7H14O2 | 130.19 g/mol
Quantity:
5 mL
100 mL
Packaging:
Glass bottle
Catalog number 150481000
Price (JPY)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS108-64-5
IUPAC Nameethyl 3-methylbutanoate
Molecular FormulaC7H14O2
InChI KeyPPXUHEORWJQRHJ-UHFFFAOYSA-N
SMILESCCOC(=O)CC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Specific gravity(25/25°C) 0.861 to 0.865
Infrared spectrumConforms
Refractive index1.3950 to 1.3980 (20°C, 589 nm)
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only