Ethyl propionate, 99+%
Ethyl propionate, 99+%
Ethyl propionate, 99+%
Thermo Scientific Chemicals

Ethyl propionate, 99+%

CAS: 105-37-3 | C5H10O2 | 102.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 150500010
Price (USD)
62.65
Special offer
Online exclusive
Ends: 28-Dec-2026
73.30
Save 10.65
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
62.65
Special offer
Online exclusive
Ends: 28-Dec-2026
73.30
Save 10.65
Each
Chemical Identifiers
CAS105-37-3
IUPAC Nameethyl propanoate
Molecular FormulaC5H10O2
InChI KeyFKRCODPIKNYEAC-UHFFFAOYSA-N
SMILESCCOC(=O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.3830 to 1.3870 (20°C, 589 nm)
Acid value=<1 mg KOH/g
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only