Isoamyl acetate, 99%, pure, mixture of 2- and 3-Methylbutyl acetate
Isoamyl acetate, 99%, pure, mixture of 2- and 3-Methylbutyl acetate
Isoamyl acetate, 99%, pure, mixture of 2- and 3-Methylbutyl acetate
Thermo Scientific Chemicals

Isoamyl acetate, 99%, pure, mixture of 2- and 3-Methylbutyl acetate

CAS: 123-92-2 | C7H14O2 | 130.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 150660010
Price (USD)
70.65
Special offer
Online exclusive
Ends: 28-Dec-2026
83.20
Save 12.55
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
70.65
Special offer
Online exclusive
Ends: 28-Dec-2026
83.20
Save 12.55
Each
Chemical Identifiers
CAS123-92-2
IUPAC Name3-methylbutyl acetate
Molecular FormulaC7H14O2
InChI KeyMLFHJEHSLIIPHL-UHFFFAOYSA-N
SMILESCC(C)CCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.3990 to 1.4030 (20°C, 589 nm)
GC>=98.5 % (sum of isomers)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only