N,N-Diethylethylenediamine, 98+%
N,N-Diethylethylenediamine, 98+%
N,N-Diethylethylenediamine, 98+%
Thermo Scientific Chemicals

N,N-Diethylethylenediamine, 98+%

CAS: 100-36-7 | C6H16N2 | 116.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 150701000
Price (USD)
31.65
Special offer
Online exclusive
Ends: 28-Dec-2026
37.20
Save 5.55
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
31.65
Special offer
Online exclusive
Ends: 28-Dec-2026
37.20
Save 5.55
Each
Chemical Identifiers
CAS100-36-7
IUPAC Name(2-aminoethyl)diethylamine
Molecular FormulaC6H16N2
InChI KeyUDGSVBYJWHOHNN-UHFFFAOYSA-N
SMILESCCN(CC)CCN
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.0 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.4350 to 1.4370 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only