Diphenylamine, 99%, pure
Diphenylamine, 99%, pure
Diphenylamine, 99%, pure
Thermo Scientific Chemicals

Diphenylamine, 99%, pure

CAS: 122-39-4 | C12H11N | 169.23 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1507400101 kgGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 150740010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS122-39-4
IUPAC NameN-phenylaniline
Molecular FormulaC12H11N
InChI KeyDMBHHRLKUKUOEG-UHFFFAOYSA-N
SMILESN(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Color)White to yellow to beige
Appearance (Form)Crystalline powder or crystals or flakes
Infrared spectrumConforms
Melting point52°C to 55°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only