tert-Butylhydroquinone, 97%
tert-Butylhydroquinone, 97%
tert-Butylhydroquinone, 97%
Thermo Scientific Chemicals

tert-Butylhydroquinone, 97%

CAS: 1948-33-0 | C10H14O2 | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
500 g
2 kg
Packaging:
View all
Glass Bottle
Catalog number 150820050
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.10
Save 5.45
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.10
Save 5.45
Each
Chemical Identifiers
CAS1948-33-0
IUPAC Name2-tert-butylbenzene-1,4-diol
Molecular FormulaC10H14O2
InChI KeyBGNXCDMCOKJUMV-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC(O)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=96.0 %
Melting point125°C to 130°C
AppearanceMay contain black particles
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only