Tetraethylammonium chloride hydrate, 99%
Tetraethylammonium chloride hydrate, 99%
Tetraethylammonium chloride hydrate, 99%
Tetraethylammonium chloride hydrate, 99%
Thermo Scientific Chemicals

Tetraethylammonium chloride hydrate, 99%

CAS: 1015691-13-0 | C8H22ClNO | 183.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 150900250
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.90
Save 8.25
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.90
Save 8.25
Each
Chemical Identifiers
CAS56-34-8
IUPAC Nametetraethylazanium chloride
Molecular FormulaC8H20ClN
InChI KeyYMBCJWGVCUEGHA-UHFFFAOYSA-M
SMILES[Cl-].CC[N+](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White
Titration Argentometric>=98.5 % (on dry substance)
Impurity=<0.5 % Triethylamine hydrochloride
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only