Pierce™ Goat Anti-Mouse IgG Antibody Coated Plates, Clear, 96-Well
Pierce™ Goat Anti-Mouse IgG Antibody Coated Plates, Clear, 96-Well
Pierce™ Goat Anti-Mouse IgG Antibody Coated Plates, Clear, 96-Well
Thermo Scientific™
Pierce™ Goat Anti-Mouse IgG Antibody Coated Plates, Clear, 96-Well
Thermo Scientific Pierce Goat Anti-Mouse IgG Antibody Coated Plates are ideal for binding assays when available antibodies are in lowRead more
Have Questions?
Catalog NumberColor
15134Clear
Catalog number 15134
Price (USD)
281.00
Each
Color:
Clear
Request bulk or custom format
Price (USD)
281.00
Each
Thermo Scientific Pierce Goat Anti-Mouse IgG Antibody Coated Plates are ideal for binding assays when available antibodies are in low quantities or they denature and become inactive upon direct adsorption to polystyrene plates. These 96-well plates are clear for colorimetric assays.
Features of Goat Anti-Mouse IgG Antibody Coated Plates:
• Prevents antibody denaturation as a result of direct binding to polystyrene
• Unlike Protein A or Protein G Plates, these plates bind only to target species
• Antibody-binding capacity is higher than direct adsorption onto polystyrene
• Pre-blocked to reduce nonspecific binding
• Binding Capacity: ∼7 pmol IgG/well
• Activation level: 100 μL/well
Because these plates are IgG-specific, purified antibodies are not required. These plates may be used for direct, indirect, competitive or sandwich assays.
Related Products
Pierce™ Goat Anti-Mouse IgG Antibody Coated Plates, White, 96-Well
Pierce™ Goat Anti-Mouse IgG Antibody Coated Plates, Black, 96-Well
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding PropertyStandard Capacity, Affinity
Detection MethodColorimetric
Packaging5 Plate
Plate BlockingSuperBlock™ Buffer
Product TypeDetection Plate, Immunoassay, ELISA
ColorClear
MaterialPolystyrene
No. of Wells96
Product LinePierce
Surface TreatmentAntibody
Unit SizeEach
Contents & Storage
Note: Plates are activated to 100 μL and supplied pre-blocked with SuperBlock™ Blocking Buffer.
Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag and store desiccated at 4°C.