Cresol Red, pure, Indicator grade
Cresol Red, pure, Indicator grade
Cresol Red, pure, Indicator grade
Thermo Scientific Chemicals

Cresol Red, pure, Indicator grade

CAS: 1733-12-6 | C21H18O5S | 382.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 151381000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1733-12-6
IUPAC Name3,3-bis(4-hydroxy-3-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC21H18O5S
InChI KeyOBRMNDMBJQTZHV-UHFFFAOYSA-N
SMILESCC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Specific extinction E (1%/1cm)>=1500 (572 nm, in 0.01 N NaOH)
Appearance (Color)Dark green to red-brown
Infrared spectrumConforms
Solubility(0.1 % in 50 % ethanol) Clear
Visual transition intervalfrom pH 7.0 (orange) to pH 8.8 (purple)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only