Penicillin-Streptomycin (10,000 U/mL), 20 x 100mL - FAQs
Penicillin-Streptomycin (10,000 U/mL), 20 x 100mL - FAQs
View additional product information for Penicillin-Streptomycin (10,000 U/mL) - FAQs (15140163, 15140148, 15140122)
1 product FAQs found
How does Penicillin work?
Penicillin interferes with the final stages of bacterial cell wall synthesis.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.