Methyl Red, sodium salt, pure, indicator
Methyl Red, sodium salt, pure, indicator
Methyl Red, sodium salt, pure, indicator
Thermo Scientific Chemicals

Methyl Red, sodium salt, pure, indicator

CAS: 845-10-3 | C15H14N3NaO2 | 291.286 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 151410250
Price (USD)
42.60
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
42.60
Each
Chemical Identifiers
CAS845-10-3
IUPAC Namesodium 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoate
Molecular FormulaC15H14N3NaO2
InChI KeyGNTPCYMJCJNRQB-UHFFFAOYSA-M
SMILES[Na+].CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red to brown
Infrared spectrumConforms
Visual transition intervalFrom pH 4.2 (pink) to pH 6.2 (yellow)
Appearance (Form)Powder
Solubility(0.1% in water) clear to faint turbid solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only