Methyl Orange, pure, indicator
Methyl Orange, pure, indicator
Methyl Orange, pure, indicator
Thermo Scientific Chemicals

Methyl Orange, pure, indicator

CAS: 547-58-0 | C14H14N3NaO3S | 327.334 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 151421000
Price (USD)
42.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
42.00
Each
Chemical Identifiers
CAS547-58-0
IUPAC Name4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonate
Molecular FormulaC14H14N3O3S
InChI KeyIETWCRRCPURZOC-UHFFFAOYSA-M
SMILESCN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Visual transition intervalfrom pH 3.2 (pink or red)to pH 4.4 (yellow-orange)
Infrared spectrumConforms
Appearance (Color)Orange
Solubility(0.1 % in water) Clear
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only