Thermo Scientific Pierce Copper Coated High Binding Capacity (HBC) Plates use an exclusive coating process to increase the amount of histidine-tagged protein that will bind to the plate surface.
Have Questions?
Change view
Catalog Number
Color
Format
Detection Method
15148
Black
Standard
Fluorescence
15146
Clear
Strip Plate
Colorimetric
15143
Clear
Standard
Colorimetric
15147
White
Standard
Chemiluminescence
4 Options
Catalog number 15148
Price (USD)
395.65
Online exclusive
421.00
Save 25.35
Each
Color:
Black
Format:
Standard
Detection Method:
Fluorescence
Request bulk or custom format
Price (USD)
395.65
Online exclusive
421.00
Save 25.35
Each
Pierce Copper Coated Plates are ideal for analyzing polyhistidine-tagged fusion proteins by ELISA-based methods. Proteins that contain a succession of several histidine residues at the amino or carboxyl terminus have a strong binding affinity for metals such as copper. Although copper is less discriminating in its binding specificity than Nickel, the higher binding capacity of these plates is ideal for high throughput screening applications that need improved sensitivity and greater dynamic range.
Thermo Scientific Pierce Copper Coated High Binding Capacity (HBC) Plates use an exclusive coating process to increase the amount of histidine-tagged protein that will bind to the plate surface.
Features of Copper Coated High Binding Capacity Plates include: • Quantitate previously undetectable histidine-tagged proteins • Wider dynamic range with four-fold greater capacity than regular nickel chelate-coated plate • Copper chelate provides greater binding capacity than nickel • The detection limit is 0.1 pg of polyhistidine tagged fusion protein per well • Pre-blocked for the ultimate convenience
Note: Copper coated plates are for use with purified His-tagged proteins. Typical cell lysates contain non-his-tagged proteins that may bind to copper, which can reduce binding capacity and signal-to-noise ratios. For cell lysates, use Thermo Scientific Pierce Nickel Coated Plates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding PropertyHigh Capacity, Affinity
Detection MethodFluorescence
FormatStandard
Packaging5 Plates
Plate BlockingBlocker™ BSA Buffer
Product TypeMicroplate
Quantity5 plates
ColorBlack
MaterialPolystyrene
No. of Wells96
Product LinePierce
Surface TreatmentDivalent Copper, Metal Chelate
Unit SizeEach
Contents & Storage
Store at 4°C in unopened pouches. Once opened, store unused plates in a resealable bag with desiccant at 4°C.
Frequently asked questions (FAQs)
Do Pierce Copper Coated High Capacity Plates, White, 96-Well (Cat. No. 15147) and Pierce Copper Coated High Capacity Plates, Black, 96-Well (Cat. No. 15148) have clear bottoms to be read from below?
Pierce Copper Coated High Capacity Plates, White, 96-Well (Cat. No. 15147) and Pierce Copper Coated High Capacity Plates, Black, 96-Well (Cat. No. 15148) can only be read from the top as they have opaque bottoms.