4-(4-Nitrophenylazo)resorcinol, 90%
4-(4-Nitrophenylazo)resorcinol, 90%
4-(4-Nitrophenylazo)resorcinol, 90%
Thermo Scientific Chemicals

4-(4-Nitrophenylazo)resorcinol, 90%

CAS: 74-39-5 | C12H9N3O4 | 259.22 g/mol
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 151510250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS74-39-5
IUPAC Name3-hydroxy-4-[2-(4-nitrophenyl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
Molecular FormulaC12H9N3O4
InChI KeyBIFFKKLAAIGZBV-UHFFFAOYSA-N
SMILESOC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange-red to red
HPLC>=88 %
Appearance (Form)Powder
Specific extinction E (1%/1cm)>=1400 (554 to 559 nm, 1N NaOH)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only