4-Nitrophenethyl bromide, 98%
4-Nitrophenethyl bromide, 98%
4-Nitrophenethyl bromide, 98%
Thermo Scientific Chemicals

4-Nitrophenethyl bromide, 98%

CAS: 5339-26-4 | C8H8BrNO2 | 230.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 151620050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5339-26-4
IUPAC Name1-(2-bromoethyl)-4-nitrobenzene
Molecular FormulaC8H8BrNO2
InChI KeyNTURQZFFJDCTMZ-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(CCBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystalline powder or crystals
GC>=97.5 %
Infrared spectrumConforms
Melting point67°C to 70°C
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only